Product Details

SNP ID
rs4165124
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:20500205 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TATAGGTCCTCTTGCCCCTGGCCAA[C/T]GTTGCGCTAATTGACTATACTCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Eif2b5 PubMed Links

Gene Details

Gene
Eif2b5
Gene Name
eukaryotic translation initiation factor 2B, subunit 5 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172265.2 301 Silent Mutation NP_758469.1

View Full Product Details