Product Details
- SNP ID
-
rs13476912
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:170425455 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCATTCTTCCAGGTCTCACTGC[C/T]TGCTTGTTGGTTTAATGGAGAAGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Bcas1
PubMed Links
Gene Details
- Gene
- Bcas1
- Gene Name
- breast carcinoma amplified sequence 1
View Full Product Details