Product Details
- SNP ID
-
rs13483763
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:56759724 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTGTCAGTACATCTCAATAGCTG[C/T]AAGCAAAGTTTCTGTCTTATTGATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fhl1
PubMed Links
Gene Details
- Gene
- Fhl1
- Gene Name
- four and a half LIM domains 1
View Full Product Details