Product Details
- SNP ID
-
rs3687432
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:59215594 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCGCAGGCCACCTGGAGCACCAC[G/T]CGTCCAGCAAGGTGTTCCACCTTCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Als2
PubMed Links
Gene Details
- Gene
- Als2
- Gene Name
- amyotrophic lateral sclerosis 2 (juvenile)
There are no transcripts associated with this gene.
View Full Product Details