Product Details
- SNP ID
-
rs13478537
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:132419098 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGGGCCTAACCTTGAGGGTCGCC[A/G]ACAAGCACCCATAGCTGTCACAATG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Auts2
PubMed Links
Gene Details
- Gene
- Auts2
- Gene Name
- autism susceptibility candidate 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177047.3 |
|
Intron |
|
|
NP_796021.2 |
View Full Product Details