Product Details
- SNP ID
-
rs13480844
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5372351 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACACAGGCTTAGAACTCCATGAC[A/G]CTCACTAGACAGTATGTTAGCAGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Znrf3
PubMed Links
Gene Details
- Gene
- Znrf3
- Gene Name
- zinc and ring finger 3
View Full Product Details