Product Details

SNP ID
rs13481657
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:112503280 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACTGATGGTAGAAGGCAGAGTTGT[C/T]GACGTTCAGCTCCAAGTCCGTTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A530016L24Rik PubMed Links

Gene Details

Gene
A530016L24Rik
Gene Name
RIKEN cDNA A530016L24 gene
There are no transcripts associated with this gene.

Gene
Tmem179
Gene Name
transmembrane protein 179
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178915.3 596 Missense Mutation NP_849246.2

View Full Product Details