Product Details
- SNP ID
-
rs3023747
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:20961338 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCCTGTCTCCAGTGGGTGTTCT[C/T]AAACTGCACTTATCGGAACTCTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rnf125
PubMed Links
Gene Details
- Gene
- Rnf125
- Gene Name
- ring finger protein 125
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_026301.2 |
|
Intron |
|
|
NP_080577.1 |
View Full Product Details