Product Details

SNP ID
rs13481057
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:62043364 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGGCAATGATGGCAGTGGAT[C/T]GTGGAAAGGCCCAAGATGAAGAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Specc1 PubMed Links

Gene Details

Gene
Specc1
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029936.3 Intron NP_001025107.1
NM_001281818.1 Intron NP_001268747.1

View Full Product Details