Product Details

SNP ID
rs13477648
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:36483036 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGATTCTGAATGACGTGAATG[A/T]GAAGTGAGGTAGAGCAAGGATTATC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Lingo2 PubMed Links

Gene Details

Gene
Lingo2
Gene Name
leucine rich repeat and Ig domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165999.1 Intron NP_001159471.1
NM_001166000.1 Intron NP_001159472.1
NM_001166001.1 Intron NP_001159473.1
NM_175516.4 Intron NP_780725.2

View Full Product Details