Product Details
- SNP ID
-
rs13477631
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:32601569 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCTTTCAGGTAAAGAAGAACAGA[C/T]CATACACTGCTGGGTCCCTGAGTAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gja10
PubMed Links
Gene Details
- Gene
- Gja10
- Gene Name
- gap junction protein, alpha 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010289.2 |
814 |
Missense Mutation |
|
|
NP_034419.2 |
View Full Product Details