Product Details

SNP ID
rs3091174
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:32696187 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GACCTCCTTCTCCATCCCGCTCATC[A/G]TCTCTCTGCCAACGTTTTGTGAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Sema5a PubMed Links

Gene Details

Gene
Sema5a
Gene Name
sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009154.2 10655 UTR 3 NP_033180.2

View Full Product Details