Product Details

SNP ID
rs29358779
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:40728912 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTTCATCCAGAGTCCCCTGGG[C/T]GACATCTGAGGGGAGACAATCACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Mettl24 PubMed Links

Gene Details

Gene
Mettl24
Gene Name
methyltransferase like 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177793.3 Intron NP_808461.1

View Full Product Details