Product Details
- SNP ID
-
rs6217738
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72672817 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAAAGCAGTATTTATAGTGGTAT[A/G]TTAAACTGCATATTTGTATTCAGGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ube2g1
PubMed Links
Gene Details
- Gene
- Ube2g1
- Gene Name
- ubiquitin-conjugating enzyme E2G 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025985.4 |
|
Intron |
|
|
NP_080261.2 |
View Full Product Details