Product Details
- SNP ID
-
rs13476878
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:160769245 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACCCATGCTTGGTACCCAAAGCC[C/T]TTCCCTGGAGTGCCTACGCTATGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Plcg1
PubMed Links
Gene Details
- Gene
- Plcg1
- Gene Name
- phospholipase C, gamma 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021280.3 |
|
Intron |
|
|
NP_067255.2 |
- Gene
- Zhx3
- Gene Name
- zinc fingers and homeoboxes 3
There are no transcripts associated with this gene.
View Full Product Details