Product Details
- SNP ID
-
rs13476276
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:182891165 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGAGGGCTGATGCCGCCAGTCCC[C/T]GACAAAAGCCATGCCCAGGCATCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Susd4
PubMed Links
Gene Details
- Gene
- Susd4
- Gene Name
- sushi domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144796.4 |
|
Intron |
|
|
NP_659045.2 |
View Full Product Details