Product Details

SNP ID
rs4225104
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:24483299 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGACAGGCGGGCAGGACTCGCCAGA[A/G]AGGCCCCAGACCTCTAACGTGTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Agap3 PubMed Links

Gene Details

Gene
Agap3
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256431.1 Intron NP_001243360.1
NM_139153.2 Intron NP_631892.2

View Full Product Details