Product Details

SNP ID
rs13476520
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:58009358 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGGACTAATTATTCTAATAAAAGT[A/C]TCACCCAGTTCCTTCTCAGTGGACC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Galnt5 PubMed Links

Gene Details

Gene
Galnt5
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172855.3 Intron NP_766443.2

View Full Product Details