Product Details
- SNP ID
-
rs6219100
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:96108732 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATTCCACTGGGTGTATCTGCTC[A/C]AGCAAAGCTGAGGGAAGAGTTTACT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Sv2c
PubMed Links
Gene Details
- Gene
- Sv2c
- Gene Name
- synaptic vesicle glycoprotein 2c
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_029210.1 |
|
Intron |
|
|
NP_083486.1 |
View Full Product Details