Product Details

SNP ID
rs13475818
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:38036741 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTAGACCGGCCTTGACTCTTCTGC[A/G]TCTGTCCCTCAGTGCTAGGATTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Eif5b PubMed Links

Gene Details

Gene
Eif5b
Gene Name
eukaryotic translation initiation factor 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198303.2 Intron NP_938045.2

View Full Product Details