Product Details

SNP ID
rs13482854
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:7136976 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCAAGCATCTCTTTGTCCTC[A/G]TGGGACCAACATGGCTGGGTGTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Rps6ka2 PubMed Links

Gene Details

Gene
Rps6ka2
Gene Name
ribosomal protein S6 kinase, polypeptide 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011299.4 Intron NP_035429.1

View Full Product Details