Product Details
- SNP ID
-
rs6288723
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:132805583 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTACCCTCCACCGCCACTCATCAA[C/T]AATTTGGGATGACTCCGATCTGGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Trmt6
PubMed Links
Gene Details
- Gene
- Trmt6
- Gene Name
- tRNA methyltransferase 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175113.3 |
1452 |
Silent Mutation |
|
|
NP_780322.2 |
View Full Product Details