Product Details
- SNP ID
-
rs6302276
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:39169435 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCTGTGTGTTAGCACTCCAGAGG[C/T]AGCGGCAGGAGGATGTGGAGTTTGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Arhgap26
PubMed Links
Gene Details
- Gene
- Arhgap26
- Gene Name
- Rho GTPase activating protein 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175164.4 |
|
Intron |
|
|
NP_780373.3 |
View Full Product Details