Product Details
- SNP ID
-
rs13477379
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:122837708 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATGTTAGAGTAGAAACATCTGG[A/G]CTTTCCTTGGGAAAGTGCTCTGTAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pde5a
PubMed Links
Gene Details
- Gene
- Pde5a
- Gene Name
- phosphodiesterase 5A, cGMP-specific
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153422.2 |
|
Intron |
|
|
NP_700471.2 |
View Full Product Details