Product Details

SNP ID
rs13477839
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:91342519 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCCACCCATGGGCTCCGGGGC[G/T]TCAATAGCGTATTTCAGTTCCCATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Elavl2 PubMed Links

Gene Details

Gene
Elavl2
Gene Name
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177883.1 Intron NP_001171354.1
NM_010486.2 Intron NP_034616.1
NM_207685.1 Intron NP_997568.1
NM_207686.1 Intron NP_997569.1

View Full Product Details