Product Details
- SNP ID
-
rs13483415
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:67472992 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGCACAGTCTGCGCCTGCTCAG[C/T]ACCGAATGGGGGCTGCCTGGCCTCG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Slmo1
PubMed Links
Gene Details
- Gene
- Slmo1
- Gene Name
- slowmo homolog 1 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144867.2 |
244 |
Silent Mutation |
|
|
NP_659116.1 |
View Full Product Details