Product Details
- SNP ID
-
rs4195412
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:63792473 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAATGCATTGCTTTGTCAAGTTC[G/A]ACTTATTCTCATTTCTCATAATTGA
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Epha3
PubMed Links
Gene Details
- Gene
- Epha3
- Gene Name
- Eph receptor A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010140.3 |
|
Intron |
|
|
NP_034270.1 |
View Full Product Details