Product Details
- SNP ID
-
rs13477737
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:62346092 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGCACTGCAGTGGCAAACAGCAC[A/T]GAGGACGTGCCTGTGGTAGCTGGGG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Fkbp15
PubMed Links
Gene Details
- Gene
- Fkbp15
- Gene Name
- FK506 binding protein 15
View Full Product Details