Product Details

SNP ID
rs13477456
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:142249507 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCCCAGAGCCACAAGAAAAGG[C/T]CCCCTGCCAAAAAGAAAGAGACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pdlim5 PubMed Links

Gene Details

Gene
Pdlim5
Gene Name
PDZ and LIM domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190852.1 1494 Silent Mutation NP_001177781.1
NM_001190853.1 1494 Intron NP_001177782.1
NM_001190854.1 1494 Silent Mutation NP_001177783.1
NM_001190855.1 1494 Silent Mutation NP_001177784.1
NM_001190856.1 1494 Silent Mutation NP_001177785.1
NM_001190857.1 1494 Intron NP_001177786.1
NM_019808.3 1494 Silent Mutation NP_062782.2
NM_019809.3 1494 Intron NP_062783.2
NM_022554.3 1494 Intron NP_072048.2

View Full Product Details