Product Details

SNP ID
rs4207452
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:77429184 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTTCAAAAGTGGCAGAAATTCAAA[A/G]GTTTCCATTTGGTTTATTGGAATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mir99ahg PubMed Links

Gene Details

Gene
Mir99ahg
Gene Name
Mir99a and Mirlet7c-1 host gene (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details