Product Details

SNP ID
rs13478105
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:8291659 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GATTTGGTAACAAAGATCGCATTGT[C/T]TTACATTTCAAGACTCATTTGGCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Adam22 PubMed Links

Gene Details

Gene
Adam22
Gene Name
a disintegrin and metallopeptidase domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007220.2 Intron NP_001007221.1
NM_001007221.2 Intron NP_001007222.1
NM_001098225.1 Intron NP_001091695.1

View Full Product Details