Product Details

SNP ID
rs13459183
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:28638445 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACAACATGGGGATGGTCGGGACACA[C/T]GGGCTGGAAACTTCGCATGCGGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Tnik PubMed Links

Gene Details

Gene
Tnik
Gene Name
TRAF2 and NCK interacting kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163007.1 3078 Silent Mutation NP_001156479.1
NM_001163008.1 3078 Silent Mutation NP_001156480.1
NM_001163009.1 3078 Silent Mutation NP_001156481.1
NM_026910.1 3078 Silent Mutation NP_081186.1

View Full Product Details