Product Details

SNP ID
rs6319490
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:84906212 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTTCAGGGGAGAGAGCCAGTGTTC[C/T]ATCCGCCTCATGCTCTCATCTGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Myo19 PubMed Links

Gene Details

Gene
Myo19
Gene Name
myosin XIX
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025414.3 Intron NP_079690.2
Gene
Znhit3
Gene Name
zinc finger, HIT type 3
There are no transcripts associated with this gene.

View Full Product Details