Product Details
- SNP ID
-
rs6260898
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:12585106 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACTTTCTGTGGCTGCTGTCATCT[C/G]TATTTGAGTGGAAGTACCTCAAGTA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Ero1lb
PubMed Links
Gene Details
- Gene
- Ero1lb
- Gene Name
- ERO1-like beta (S. cerevisiae)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_026184.2 |
|
Intron |
|
|
NP_080460.2 |
View Full Product Details