Product Details

SNP ID
rs13481018
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:50383262 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TAACTGGTGAAGCAGATGTTGAGTT[C/T]GCAACCCATGAAGATGCTGTTGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Hnrnph1 PubMed Links

Gene Details

Gene
Hnrnph1
Gene Name
heterogeneous nuclear ribonucleoprotein H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021510.2 1129 Silent Mutation NP_067485.1
Gene
Rufy1
Gene Name
RUN and FYVE domain containing 1
There are no transcripts associated with this gene.

View Full Product Details