Product Details

SNP ID
rs3716162
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:59036258 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTCAGTTCCCTCTGTTTTAGCCCTC[G/A]ACCTGGTGCTACTCCAGGGAATAGG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
AI481877 PubMed Links

Gene Details

Gene
AI481877
Gene Name
expressed sequence AI481877
There are no transcripts associated with this gene.

Gene
Gng10
Gene Name
guanine nucleotide binding protein (G protein), gamma 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025277.3 Intron NP_079553.1

View Full Product Details