Product Details
- SNP ID
-
rs13477199
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:75071754 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTCCTTGATATCGTTCCTTTATA[A/G]GTTTTTTCCTAATGGTGACTTTTTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Zbbx
PubMed Links
Gene Details
- Gene
- Zbbx
- Gene Name
- zinc finger, B-box domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172515.2 |
1596 |
Silent Mutation |
|
|
NP_766103.2 |
View Full Product Details