Product Details

SNP ID
rs3022960
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:65329813 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAACGGAAGAATGAACACTTGATA[C/T]GCCTTGTCTCCATGTTTCAAGCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A730090N16Rik PubMed Links

Gene Details

Gene
A730090N16Rik
Gene Name
RIKEN cDNA A730090N16 gene
There are no transcripts associated with this gene.

Gene
Kcnab1
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 1
There are no transcripts associated with this gene.

View Full Product Details