Product Details
- SNP ID
-
rs13475799
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:33754441 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTCATGAACACGGGCTCATTTG[C/G]CAATGACAAAATTGCGTGCTTTCAT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Bag2
PubMed Links
Gene Details
- Gene
- Bag2
- Gene Name
- BCL2-associated athanogene 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145392.2 |
|
Intron |
|
|
NP_663367.1 |
- Gene
- Zfp451
- Gene Name
- zinc finger protein 451
There are no transcripts associated with this gene.
View Full Product Details