Product Details

SNP ID
rs3676330
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:128210048 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGTCAGACAAGTCCAAACTG[A/T]ATATTCCGCATACATATTCAACTGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Gls2 PubMed Links

Gene Details

Gene
Gls2
Gene Name
glutaminase 2 (liver, mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033264.3 1430 Intron NP_001028436.2
NM_001285777.1 1430 Intron NP_001272706.1
NM_001285779.1 1430 Intron NP_001272708.1
Gene
Spryd4
Gene Name
SPRY domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025716.2 1430 UTR 3 NP_079992.1

View Full Product Details