Product Details

SNP ID
rs13483308
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:38268874 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTCTACCCTGTGCTTCTGAAGCTGA[A/G]GCCCCACTGGTGGCTAGGTCTAGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
0610009O20Rik PubMed Links

Gene Details

Gene
0610009O20Rik
Gene Name
RIKEN cDNA 0610009O20 gene
There are no transcripts associated with this gene.

Gene
Pcdh12
Gene Name
protocadherin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017378.2 3805 Silent Mutation NP_059074.2

View Full Product Details