Product Details

SNP ID
rs13478578
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:145218528 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATGTTAACCGGAAGCAGAAATCGAA[C/T]GGCGAACGAGGCCACAGATGTGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Zkscan5 PubMed Links

Gene Details

Gene
Zkscan5
Gene Name
zinc finger with KRAB and SCAN domains 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167944.1 1744 Silent Mutation NP_001161416.1
NM_016683.2 1744 Silent Mutation NP_057892.2

View Full Product Details