Product Details

SNP ID
rs13480767
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:112399386 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTAATCAAAGCAAGAACAAACCTCG[A/G]ATGTAGTCCTGCTGAAGGAACAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Kcnc2 PubMed Links

Gene Details

Gene
Kcnc2
Gene Name
potassium voltage gated channel, Shaw-related subfamily, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025581.1 Intron NP_001020752.1

View Full Product Details