Product Details
- SNP ID
-
rs13481609
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:99263253 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTACAAACACCAACTCATCTGAAT[A/G]CCAAACATCGTTCCATAATTGAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Foxn3
PubMed Links
Gene Details
- Gene
- Foxn3
- Gene Name
- forkhead box N3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183186.2 |
|
Intron |
|
|
NP_899009.2 |
View Full Product Details