Product Details

SNP ID
rs13478544
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:135965541 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCCGAAGACCCCGCCCCGTA[C/G]CCTGGCACCGCCCCCGCTGCCCCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Ssc4d PubMed Links

Gene Details

Gene
Ssc4d
Gene Name
scavenger receptor cysteine rich family, 4 domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160366.1 Intron NP_001153838.1

View Full Product Details