Product Details

SNP ID
rs13477723
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:57203906 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCACACAGGTAGAAGGGACAGCGGG[A/G]ACTGACCCTGGCTGGCTGGCTGGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ptpn3 PubMed Links

Gene Details

Gene
Ptpn3
Gene Name
protein tyrosine phosphatase, non-receptor type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011207.2 Intron NP_035337.2

View Full Product Details