Product Details

SNP ID
rs6305730
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:67856295 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGAAAGCAGTGTTATGTCTC[A/G]TGTGTATGTACATTGGCTGTACATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3galt1 PubMed Links

Gene Details

Gene
B3galt1
Gene Name
UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020283.4 Intron NP_064679.2

View Full Product Details