Product Details
- SNP ID
-
rs4229759
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:55506933 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAGGAGGTTCAGGCCAGGGACAT[G/A]AAAGTGGTAGGCAAGAGCTGGAGGC
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mir6945
PubMed Links
Gene Details
- Gene
- Mir6945
- Gene Name
- microRNA 6945
There are no transcripts associated with this gene.
- Gene
- Pdlim7
- Gene Name
- PDZ and LIM domain 7
View Full Product Details