Product Details
- SNP ID
-
rs3668963
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:79352330 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCATCTTCAAAGCAAAAAGATG[G/A]CAATGCCATTCCTGGAAATAGCCCC
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cerkl
PubMed Links
Gene Details
- Gene
- Cerkl
- Gene Name
- ceramide kinase-like
View Full Product Details