Product Details
- SNP ID
-
rs3660676
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:24860398 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTGCCTTCAAATGCTGGATGGGT[C/T]CTTGTCTGGAGATGGACACTGGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fhod3
PubMed Links
Gene Details
- Gene
- Fhod3
- Gene Name
- formin homology 2 domain containing 3
There are no transcripts associated with this gene.
View Full Product Details